Assay Details
Target Gene Details
Entrez Gene ID: | 5553 |
Gene Name: | proteoglycan 2, pro eosinophil major basic protein |
Gene Aliases: |
BMPG, MBP, MBP1, proMBP |
Location: |
Chr.11:57386787-57390657 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PRG2 | NM_001243245.2 | 6 | 797 | NP_001230174.1 |
| NM_001302926.1 | 6 | 826 | NP_001289855.1 | |
| NM_001302927.1 | 6 | 943 | NP_001289856.1 | |
| NM_002728.5 | 6 | 830 | NP_002719.3 | |
| BC005929.1 | AAH05929.1 | |||
| BQ445845.1 | ||||
| BX395670.2 | 1 | 559 | ||
| BX397850.2 | 1 | 595 | ||
| BX454677.2 | 1 | 605 | ||
| M35670.1 | AAA35965.1 | |||
| M36805.1 | AAA36203.1 | |||
| X14088.1 | CAA32250.1 | |||
| X65787.1 | CAA46670.1 | |||
| Y00809.1 | CAA68751.1 | |||
| Z26248.1 | CAA81207.1 |
More Information
Set Membership: |
|

Genomic Map