Assay Details
Target Gene Details
Entrez Gene ID: | 5196 |
Gene Name: | platelet factor 4 |
Gene Aliases: |
CXCL4, PF-4, SCYB4 |
Location: |
Chr.4:73980825-73982124 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PF4 | NM_002619.3 | 1 | 185 | NP_002610.1 |
| AJ705588.1 | 1 | 59 | ||
| BC093965.1 | AAH93965.1 | |||
| BC112093.1 | AAI12094.1 | |||
| CR407677.1 | CAG28605.1 | |||
| M25897.1 | AAA60066.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1006029 | Chr.4:73928205 - 74120947 on Build GRCh38 | Gain |
|
| esv3575747 | Chr.4:73914851 - 74124725 on Build GRCh38 | Gain |
|
| nsv1001725 | Chr.4:73488333 - 73995207 on Build GRCh38 | Gain |
|
| nsv525980 | Chr.4:73865809 - 74556070 on Build GRCh38 | Gain |
|
| nsv1004768 | Chr.4:73908655 - 74243862 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map