Assay Details
Target Gene Details
Entrez Gene ID: | 2323 |
Gene Name: | fms related tyrosine kinase 3 ligand |
Gene Aliases: |
FL, FLT3L |
Location: |
Chr.19:49474172-49487038 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FLT3LG | NM_001204502.1 | 6 | 532 | NP_001191431.1 |
| NM_001204503.1 | 5 | 501 | NP_001191432.1 | |
| NM_001278637.1 | 6 | 560 | NP_001265566.1 | |
| NM_001278638.1 | 6 | 549 | NP_001265567.1 | |
| NM_001459.3 | 6 | 532 | NP_001450.2 | |
| XM_005258680.4 | XP_005258737.3 | |||
| XM_005258681.4 | XP_005258738.3 | |||
| XM_005258682.4 | XP_005258739.3 | |||
| XM_005258683.4 | XP_005258740.3 | |||
| XM_006723116.3 | XP_006723179.2 | |||
| XM_011526675.2 | 6 | 568 | XP_011524977.1 | |
| XM_011526676.2 | 6 | 533 | XP_011524978.1 | |
| XM_011526677.2 | 5 | 522 | XP_011524979.1 | |
| XM_011526678.2 | XP_011524980.1 | |||
| XM_011526680.2 | XP_011524982.1 | |||
| XM_011526682.2 | 5 | 539 | XP_011524984.1 | |
| XM_017026532.1 | XP_016882021.1 | |||
| XM_017026533.1 | 5 | 458 | XP_016882022.1 | |
| XM_017026534.1 | 5 | 431 | XP_016882023.1 | |
| XM_017026535.1 | XP_016882024.1 | |||
| AI792400.1 | ||||
| AK301136.1 | 5 | 493 | ||
| AK308939.1 | ||||
| BC006331.2 | 6 | 522 | AAH06331.1 | |
| BC011914.2 | 6 | 522 | AAH11914.1 | |
| BC028001.1 | 6 | 533 | ||
| BC126293.1 | 6 | 489 | ||
| BC136464.1 | 6 | 489 | ||
| BC144129.1 | 6 | 489 | ||
| DB490139.2 | ||||
| U03858.1 | 5 | 505 | AAA19825.1 | |
| U04806.1 | 6 | 514 | AAA17999.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv9739 | Chr.19:47901840 - 50143063 on Build GRCh38 | Gain+Loss |
|
| nsv833862 | Chr.19:49448789 - 49570676 on Build GRCh38 | Loss |
|
| nsv953599 | Chr.19:49386444 - 49506743 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map