Assay Details
Target Gene Details
Entrez Gene ID: | 105379054 |
Gene Name: | uncharacterized LOC105379054 |
Gene Aliases: |
- |
Location: |
Chr.5:83531352-83562314 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC105379054 | NR_136215.1 | |||
| DB446569.1 |
Target Gene Details
Entrez Gene ID: | 1462 |
Gene Name: | versican |
Gene Aliases: |
CSPG2, ERVR, GHAP, PG-M, WGN, WGN1 |
Location: |
Chr.5:83471674-83582303 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| VCAN | NM_001126336.2 | NP_001119808.1 | ||
| NM_001164097.1 | 7 | 3994 | NP_001157569.1 | |
| NM_001164098.1 | NP_001157570.1 | |||
| NM_004385.4 | 8 | 6955 | NP_004376.2 | |
| AB209491.1 | 8 | 6838 | BAD92728.1 | |
| AK296285.1 | ||||
| AK299770.1 | 3 | 2912 | ||
| AK307642.1 | 7 | 3895 | ||
| BX641036.1 | 1 | 3989 | CAE46022.1 | |
| D32039.1 | BAA06801.1 | |||
| U16306.1 | 8 | 6865 | AAA65018.1 | |
| U26555.1 | AAA67565.1 | |||
| X15998.1 | 7 | 3904 | CAA34128.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv830367 | Chr.5:83531833 - 83742562 on Build GRCh38 | Gain+Loss |
|
| nsv1015852 | Chr.5:83404850 - 84499122 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map