Assay Details
Target Gene Details
Entrez Gene ID: | 83479 |
Gene Name: | DEAD-box helicase 59 |
Gene Aliases: |
OFD5, ZNHIT5 |
Location: |
Chr.1:200640807-200669998 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DDX59 | NM_001031725.5 | 3 | 1130 | NP_001026895.2 |
| NM_001320181.1 | 3 | 1130 | NP_001307110.1 | |
| NM_001320182.1 | 2 | 898 | NP_001307111.1 | |
| XM_005245519.1 | 3 | 1039 | XP_005245576.1 | |
| XM_005245521.2 | 3 | 1039 | XP_005245578.1 | |
| XM_006711562.1 | 3 | 1039 | XP_006711625.1 | |
| XM_011510035.2 | 3 | 1063 | XP_011508337.1 | |
| XM_017002431.1 | 3 | 1818 | XP_016857920.1 | |
| XM_017002432.1 | 3 | 1303 | XP_016857921.1 | |
| AK299219.1 | 3 | 1001 | ||
| AL136611.1 | 3 | 1043 | CAB66546.1 | |
| BC014183.2 | 3 | 1063 | AAH14183.1 | |
| BC041801.1 | 3 | 1121 | AAH41801.1 |
More Information
Set Membership: |
|

Genomic Map