Assay Details
Target Gene Details
Entrez Gene ID: | 9229 |
Gene Name: | DLG associated protein 1 |
Gene Aliases: |
DAP-1, DAP-1-ALPHA, DAP-1-BETA, DAP1, DLGAP1A, DLGAP1B, GKAP, SAPAP1 |
Location: |
Chr.18:3496032-4455307 on Build GRCh38 |
Assay Gene Location: | Within Exon 19 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DLGAP1 | NM_001003809.2 | NP_001003809.1 | ||
| NM_001242761.1 | 12 | 3211 | NP_001229690.1 | |
| NM_001242762.1 | NP_001229691.1 | |||
| NM_001242763.1 | NP_001229692.1 | |||
| NM_001242764.1 | NP_001229693.1 | |||
| NM_001242765.1 | 9 | 1854 | NP_001229694.1 | |
| NM_001242766.1 | NP_001229695.1 | |||
| NM_001308390.1 | NP_001295319.1 | |||
| NM_004746.3 | NP_004737.2 | |||
| XM_005258171.2 | XP_005258228.1 | |||
| XM_005258172.1 | XP_005258229.1 | |||
| XM_005258173.3 | XP_005258230.2 | |||
| XM_005258174.4 | XP_005258231.1 | |||
| XM_006722367.3 | XP_006722430.1 | |||
| XM_017026080.1 | XP_016881569.1 | |||
| XM_017026081.1 | XP_016881570.1 | |||
| XM_017026082.1 | 10 | 2029 | XP_016881571.1 | |
| XM_017026083.1 | XP_016881572.1 | |||
| XM_017026084.1 | 9 | 2096 | XP_016881573.1 | |
| XM_017026085.1 | 9 | 2089 | XP_016881574.1 | |
| AB000276.1 | BAA23257.1 | |||
| AB000277.1 | BAA23258.1 | |||
| AK294717.1 | ||||
| AK294747.1 | ||||
| AK294777.1 | ||||
| AK299880.1 | ||||
| AK304101.1 | 12 | 3193 | ||
| AK316099.1 | 12 | 3211 | ||
| BC136453.1 | ||||
| BC136454.1 | ||||
| U67988.1 | 10 | 1801 | AAC51119.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1059696 | Chr.18:3494893 - 3572306 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map