Assay Details
Target Gene Details
Entrez Gene ID: | 23586 |
Gene Name: | DEXD/H-box helicase 58 |
Gene Aliases: |
RIG-I, RIGI, RLR-1, SGMRT2 |
Location: |
Chr.9:32455302-32526324 on Build GRCh38 |
Assay Gene Location: | Within Exon 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DDX58 | NM_014314.3 | 18 | 3901 | NP_055129.2 |
| AK023661.1 | 11 | 2576 | ||
| AL137608.1 | 8 | 2162 | CAB70840.1 | |
| BC132786.1 | 18 | 3849 | ||
| BC136610.1 | 18 | 3849 |
More Information
Set Membership: |
|

Genomic Map