Assay Details
Target Gene Details
Entrez Gene ID: | 8091 |
Gene Name: | high mobility group AT-hook 2 |
Gene Aliases: |
BABL, HMGI-C, HMGIC, LIPO, STQTL9 |
Location: |
Chr.12:65824460-65966291 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HMGA2 | NM_001300918.1 | 1 | 165 | NP_001287847.1 |
| NM_001300919.1 | 1 | 165 | NP_001287848.1 | |
| NM_003483.4 | 1 | 165 | NP_003474.1 | |
| NM_003484.1 | 1 | 165 | NP_003475.1 | |
| AY601861.1 | 1 | 165 | ||
| AY601862.1 | 1 | 165 | ||
| AY601863.1 | 1 | 165 | ||
| AY601864.1 | 1 | 165 | ||
| AY601865.1 | 1 | 165 | ||
| AY601866.1 | 1 | 165 | ||
| AY601867.1 | 1 | 165 | ||
| CN334368.1 | 1 | 89 | ||
| U28749.1 | 1 | 165 | AAA68613.1 | |
| X92518.1 | 1 | 680 | CAA63295.1 | |
| Z31595.1 | 1 | 166 | CAA83472.1 |
Target Gene Details
Entrez Gene ID: | 204010 |
Gene Name: | ribosomal protein SA pseudogene 52 |
Gene Aliases: |
RPSA_17_1251 |
Location: |
Chr.12:65758020-65826974 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RPSAP52 | NR_026825.2 |
More Information
Set Membership: |
|

Genomic Map