Assay Details
Target Gene Details
Entrez Gene ID: | 6749 |
Gene Name: | structure specific recognition protein 1 |
Gene Aliases: |
FACT, FACT80, T160 |
Location: |
Chr.11:57325985-57335877 on Build GRCh38 |
Assay Gene Location: | Within Exon 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SSRP1 | NM_003146.2 | 17 | 2483 | NP_003137.1 |
| XM_017018180.1 | 16 | 2700 | XP_016873669.1 | |
| AB209132.1 | 11 | 5964 | BAD92369.1 | |
| BC005116.1 | 16 | 2264 | AAH05116.1 | |
| M86737.1 | 17 | 2490 | AAA58660.1 |
More Information
Set Membership: |
|

Genomic Map