Assay Details
Target Gene Details
Entrez Gene ID: | 5037 |
Gene Name: | phosphatidylethanolamine binding protein 1 |
Gene Aliases: |
HCNP, HCNPpp, HEL-210, HEL-S-34, HEL-S-96, PBP, PEBP, PEBP-1, RKIP |
Location: |
Chr.12:118135884-118145588 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PEBP1 | NM_002567.3 | NP_002558.1 | ||
| AK226006.1 | ||||
| AK299414.1 | ||||
| AK308056.1 | ||||
| AK311927.1 | ||||
| BC008714.2 | AAH08714.1 | |||
| BC017396.1 | AAH17396.1 | |||
| BC031102.1 | AAH31102.1 | |||
| BF978793.1 | ||||
| BP347345.1 | ||||
| D16111.1 | BAA03684.1 | |||
| EU668351.1 | ||||
| EU794643.1 | ||||
| GQ472218.1 | ||||
| HM131449.1 | ||||
| S76773.1 | AAD14234.1 | |||
| X75252.1 | CAA53031.1 | |||
| X85033.1 | CAA59404.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3581916 | Chr.12:117356518 - 118228514 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map