Assay Details
Target Gene Details
Entrez Gene ID: | 11266 |
Gene Name: | dual specificity phosphatase 12 |
Gene Aliases: |
DUSP1, YVH1 |
Location: |
Chr.1:161749692-161757164 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DUSP12 | NM_007240.2 | 1 | 292 | NP_009171.1 |
| XM_005244862.2 | 1 | 368 | XP_005244919.1 | |
| AF119226.1 | 1 | 269 | AAD51134.1 | |
| BC006286.1 | 1 | 290 | AAH06286.1 | |
| BT006633.1 | 1 | 258 | AAP35279.1 | |
| ES309317.1 | 1 | 307 | ||
| HY128542.1 | 1 | 293 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv428257 | Chr.1:161436525 - 161898473 on Build GRCh38 | Loss |
|
| nsv464184 | Chr.1:161677017 - 161915562 on Build GRCh38 | Gain |
|
| esv33393 | Chr.1:161508891 - 162110537 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map