Assay Details
Target Gene Details
Entrez Gene ID: | 4915 |
Gene Name: | neurotrophic receptor tyrosine kinase 2 |
Gene Aliases: |
GP145-TrkB, TRKB, trk-B |
Location: |
Chr.9:84668368-85027070 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NTRK2 | NM_001291937.1 | 2 | 373 | NP_001278866.1 |
| NM_006180.4 | 2 | 373 | NP_006171.2 | |
| XM_005252001.2 | 2 | 463 | XP_005252058.1 | |
| XM_005252003.2 | XP_005252060.1 | |||
| XM_005252004.2 | XP_005252061.1 | |||
| XM_005252006.3 | 2 | 463 | XP_005252063.1 | |
| XM_005252007.3 | 2 | 463 | XP_005252064.1 | |
| XM_011518718.2 | 2 | 463 | XP_011517020.1 | |
| XM_011518720.2 | 2 | 463 | XP_011517022.1 | |
| XM_017014751.1 | 2 | 463 | XP_016870240.1 | |
| XM_017014752.1 | 2 | 309 | XP_016870241.1 | |
| XM_017014753.1 | 2 | 463 | XP_016870242.1 | |
| XM_017014754.1 | XP_016870243.1 | |||
| XM_017014755.1 | 2 | 309 | XP_016870244.1 | |
| XM_017014756.1 | XP_016870245.1 | |||
| XM_017014757.1 | 2 | 309 | XP_016870246.1 | |
| XM_017014758.1 | 2 | 309 | XP_016870247.1 | |
| XM_017014759.1 | XP_016870248.1 | |||
| XM_017014760.1 | 2 | 463 | XP_016870249.1 | |
| XM_017014761.1 | XP_016870250.1 | |||
| AB209118.1 | 2 | 275 | BAD92355.1 | |
| DA283111.1 | 2 | 309 | ||
| DA449367.1 | 2 | 374 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1048977 | Chr.9:84149197 - 84815576 on Build GRCh38 | Gain |
|
More Information
Additional Information:
Set Membership: |
|

Genomic Map