Assay Details
Target Gene Details
Entrez Gene ID: | 26084 |
Gene Name: | Rho guanine nucleotide exchange factor 26 |
Gene Aliases: |
CSGEF, HMFN1864, SGEF |
Location: |
Chr.3:154121003-154257827 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ARHGEF26 | NM_001251962.1 | 2 | 817 | NP_001238891.1 |
| NM_001251963.1 | 2 | 744 | NP_001238892.1 | |
| NM_015595.3 | 2 | 744 | NP_056410.3 | |
| XM_011512672.1 | 2 | 744 | XP_011510974.1 | |
| AB073386.1 | 2 | 664 | BAD38637.1 | |
| AF415175.1 | 2 | 699 | AAL27001.1 | |
| AK123040.1 | 2 | 688 | ||
| AK127282.1 | 2 | 817 | ||
| AY552599.1 | 1 | 533 | AAS59842.1 | |
| BC078655.1 | 2 | 685 | AAH78655.1 |
More Information
Set Membership: |
|

Genomic Map