Assay Details
Target Gene Details
Entrez Gene ID: | 51161 |
Gene Name: | chromosome 3 open reading frame 18 |
Gene Aliases: |
G20 |
Location: |
Chr.3:50558025-50574835 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| C3orf18 | NM_001171740.2 | 5 | 1856 | NP_001165211.1 |
| NM_001171741.2 | 5 | 1651 | NP_001165212.1 | |
| NM_001171743.2 | 5 | 1635 | NP_001165214.2 | |
| NM_016210.4 | 6 | 1945 | NP_057294.2 | |
| XM_011533781.2 | 4 | 2595 | XP_011532083.1 | |
| XM_011533782.2 | 6 | 1933 | XP_011532084.1 | |
| XM_011533783.2 | 5 | 1823 | XP_011532085.1 | |
| XM_011533784.2 | 6 | 1711 | XP_011532086.1 | |
| XM_011533785.2 | 5 | 1649 | XP_011532087.1 | |
| XM_011533788.1 | 4 | 1255 | XP_011532090.1 | |
| XM_011533789.1 | 3 | 1280 | XP_011532091.1 | |
| XM_011533790.1 | 6 | 1861 | XP_011532092.1 | |
| XM_017006545.1 | 5 | 1773 | XP_016862034.1 | |
| XM_017006546.1 | 4 | 1100 | XP_016862035.1 | |
| XM_017006547.1 | 5 | 1796 | XP_016862036.1 | |
| AF188706.1 | 5 | 1616 | AAF01050.1 | |
| AK124438.1 | 5 | 1696 | ||
| AK130227.1 | 3 | 1051 | BAC85307.1 | |
| AK308074.1 | ||||
| AL833938.1 | 6 | 1904 | CAD38793.1 | |
| BC034766.1 | 5 | 1729 | AAH34766.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3584502 | Chr.3:50327055 - 50622310 on Build GRCh38 | Gain |
|
| nsv590273 | Chr.3:50536782 - 50640720 on Build GRCh38 | Loss |
|
| nsv460532 | Chr.3:50498842 - 50578618 on Build GRCh38 | Loss |
|

Genomic Map