Assay Details
Target Gene Details
Entrez Gene ID: | 159371 |
Gene Name: | solute carrier family 35 member G1 |
Gene Aliases: |
C10orf60, TMEM20 |
Location: |
Chr.10:93893951-93914265 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC35G1 | NM_001134658.1 | NP_001128130.1 | ||
| NM_153226.2 | NP_694958.1 | |||
| XM_011539371.2 | XP_011537673.1 | |||
| XM_011539373.2 | XP_011537675.1 | |||
| AK091309.1 | BAC03633.1 | |||
| BC104814.1 | AAI04815.1 | |||
| BC143283.1 | ||||
| BC143284.1 |
More Information
Set Membership: |
|

Genomic Map