Assay Details
Target Gene Details
Entrez Gene ID: | 27177 |
Gene Name: | interleukin 36, beta |
Gene Aliases: |
FIL1, FIL1-(ETA), FIL1H, FILI-(ETA), IL-1F8, IL-1H2, IL1-ETA, IL1F8, IL1H2 |
Location: |
Chr.2:113022091-113052867 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IL36B | NM_014438.4 | NP_055253.2 | ||
| AF200494.1 | AAF69250.1 |
More Information
Set Membership: |
|

Genomic Map