Assay Details
Target Gene Details
Entrez Gene ID: | 9261 |
Gene Name: | mitogen-activated protein kinase-activated protein kinase 2 |
Gene Aliases: |
MAPKAP-K2, MK-2, MK2 |
Location: |
Chr.1:206684912-206734285 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MAPKAPK2 | NM_004759.4 | 1 | 407 | NP_004750.1 |
| NM_032960.3 | 1 | 407 | NP_116584.2 | |
| XM_005273353.3 | 1 | 515 | XP_005273410.1 | |
| BC036060.1 | 1 | 407 | AAH36060.2 | |
| BC052584.1 | 1 | 390 | AAH52584.1 | |
| X75346.1 | 1 | 185 | CAA53094.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv21658 | Chr.1:206684559 - 206685603 on Build GRCh38 | Loss |
|
| nsv1068832 | Chr.1:206684654 - 206685655 on Build GRCh38 | Deletion |
|
| nsv8713 | Chr.1:206683278 - 206686418 on Build GRCh38 | Gain |
|
| nsv549069 | Chr.1:206684741 - 206685491 on Build GRCh38 | Loss |
|
| nsv952651 | Chr.1:206684156 - 206686055 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map