Assay Details
Target Gene Details
Entrez Gene ID: | 55898 |
Gene Name: | unc-45 myosin chaperone A |
Gene Aliases: |
GC-UNC45, GCUNC-45, GCUNC45, IRO039700, SMAP-1, SMAP1, UNC-45A |
Location: |
Chr.15:90929980-90954093 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| UNC45A | NM_001039675.1 | 9 | 1343 | NP_001034764.1 |
| NM_001323619.1 | 6 | 602 | NP_001310548.1 | |
| NM_001323620.1 | 6 | 1140 | NP_001310549.1 | |
| NM_001323621.1 | 6 | 947 | NP_001310550.1 | |
| NM_018671.4 | 6 | 992 | NP_061141.2 | |
| XM_005254963.3 | 6 | 712 | XP_005255020.1 | |
| XM_011521779.2 | 9 | 1588 | XP_011520081.1 | |
| XM_011521780.2 | 5 | 494 | XP_011520082.1 | |
| XM_017022407.1 | 6 | 712 | XP_016877896.1 | |
| AB014729.1 | 6 | 534 | BAB20266.1 | |
| AB014736.1 | 6 | 576 | BAB20273.1 | |
| AK000905.1 | 5 | 870 | BAA91418.1 | |
| AK125721.1 | 9 | 1343 | ||
| AK291622.1 | 6 | 547 | ||
| AK293538.1 | 6 | 716 | ||
| AK296964.1 | 6 | 569 | ||
| AK302979.1 | 6 | 609 | ||
| BC006214.1 | 6 | 575 | AAH06214.1 | |
| BC037992.1 | 6 | 575 | AAH37992.1 | |
| BC045635.1 | 9 | 1343 | AAH45635.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833090 | Chr.15:90911195 - 91098519 on Build GRCh38 | Loss |
|
| nsv515661 | Chr.15:90866779 - 90981967 on Build GRCh38 | Loss |
|
| nsv833089 | Chr.15:90859910 - 90962961 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map