Assay Details
Target Gene Details
Entrez Gene ID: | 192669 |
Gene Name: | argonaute 3, RISC catalytic component |
Gene Aliases: |
EIF2C3 |
Location: |
Chr.1:35925198-36072037 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AGO3 | NM_024852.3 | 1 | 181 | NP_079128.2 |
| NM_177422.2 | 1 | 181 | NP_803171.1 | |
| XM_005270575.3 | XP_005270632.1 | |||
| XM_017000523.1 | XP_016856012.1 | |||
| AK027796.1 | BAB55374.1 | |||
| AK292870.1 | 1 | 147 | ||
| BC066888.1 | 1 | 181 | AAH66888.1 | |
| BF438817.1 | 1 | 92 | ||
| BF979526.1 | 1 | 186 |
More Information
Set Membership: |
|

Genomic Map