Assay Details
Target Gene Details
Entrez Gene ID: | 891 |
Gene Name: | cyclin B1 |
Gene Aliases: |
CCNB |
Location: |
Chr.5:69167010-69178245 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CCNB1 | NM_031966.3 | 1 | 124 | NP_114172.1 |
| AK289431.1 | ||||
| BI253686.1 | 1 | 124 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv5694n100 | Chr.5:69081291 - 69374274 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map