Assay Details
Target Gene Details
Entrez Gene ID: | 145873 |
Gene Name: | mesoderm posterior bHLH transcription factor 2 |
Gene Aliases: |
SCDO2, bHLHc6 |
Location: |
Chr.15:89776358-89778754 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MESP2 | NM_001039958.1 | 2 | 1423 | NP_001035047.1 |
| AL360139.1 | 3 | 668 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv570388 | Chr.15:89730911 - 89794962 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map