Assay Details
Target Gene Details
Entrez Gene ID: | 6095 |
Gene Name: | RAR related orphan receptor A |
Gene Aliases: |
NR1F1, ROR1, ROR2, ROR3, RZR-ALPHA, RZRA |
Location: |
Chr.15:60488284-61229303 on Build GRCh38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RORA | NM_002943.3 | 5 | 976 | NP_002934.1 |
| NM_134260.2 | 6 | 1000 | NP_599022.1 | |
| NM_134261.2 | 5 | 830 | NP_599023.1 | |
| NM_134262.2 | 4 | 671 | NP_599024.1 | |
| XM_005254584.4 | 5 | 5291 | XP_005254641.1 | |
| XM_011521874.1 | 4 | 868 | XP_011520176.1 | |
| XM_011521875.1 | 5 | 700 | XP_011520177.1 | |
| XM_011521877.2 | 4 | 903 | XP_011520179.1 | |
| XM_011521878.1 | 6 | 742 | XP_011520180.1 | |
| XM_011521879.2 | 2 | 628 | XP_011520181.1 | |
| XM_017022466.1 | 4 | 2692 | XP_016877955.1 | |
| XM_017022467.1 | 4 | 2365 | XP_016877956.1 | |
| AB307694.1 | 5 | 745 | ||
| AL530219.3 | 4 | 699 | ||
| AL532048.3 | 4 | 670 | ||
| BC008831.2 | 5 | 753 | AAH08831.1 | |
| BC100987.2 | 4 | 621 | AAI00988.1 | |
| BC100988.1 | 4 | 621 | AAI00989.1 | |
| BC100989.1 | 4 | 621 | AAI00990.1 | |
| BC100990.2 | 4 | 621 | AAI00991.1 | |
| BG705319.1 | ||||
| HQ692818.1 | 4 | 580 | ||
| HQ709174.1 | 4 | 580 | ||
| L14611.1 | 4 | 601 | AAA02963.1 | |
| U04897.1 | 5 | 846 | AAA62658.1 | |
| U04898.1 | 6 | 933 | AAA62659.1 | |
| U04899.1 | 5 | 898 | AAA62660.1 |
Target Gene Details
Entrez Gene ID: | 101928784 |
Gene Name: | RORA antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.15:60479178-60630639 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RORA-AS1 | NR_120339.1 | |||
| NR_120340.1 | ||||
| NR_120341.1 | ||||
| NR_120342.1 | ||||
| AI028673.1 | ||||
| AW663525.1 | ||||
| BG218240.1 | ||||
| BI668481.1 | ||||
| BX115309.1 | ||||
| DA507239.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv525689 | Chr.15:60448642 - 60531218 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map