Assay Details
Target Gene Details
Entrez Gene ID: | 7837 |
Gene Name: | peroxidasin |
Gene Aliases: |
COPOA, D2S448, D2S448E, MG50, PRG2, PXN, VPO |
Location: |
Chr.2:1631887-1744852 on Build GRCh38 |
Assay Gene Location: | Within Exon 25 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PXDN | NM_012293.2 | 23 | 6658 | NP_036425.1 |
| XM_005264707.3 | 22 | 6574 | XP_005264764.1 | |
| XM_011510396.1 | 23 | 6415 | XP_011508698.1 | |
| XM_011510397.2 | 22 | 6503 | XP_011508699.1 | |
| AF200348.1 | 23 | 6617 | AAF06354.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1005443 | Chr.2:1080998 - 1747280 on Build GRCh38 | Gain |
|
| nsv521199 | Chr.2:1622620 - 1648888 on Build GRCh38 | Loss |
|
| esv3891470 | Chr.2:1005374 - 1856757 on Build GRCh38 | Gain |
|
| nsv1007175 | Chr.2:1621349 - 1672921 on Build GRCh38 | Gain |
|
| nsv1007958 | Chr.2:1615448 - 1853325 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map