Assay Details
Target Gene Details
Entrez Gene ID: | 4356 |
Gene Name: | membrane palmitoylated protein 3 |
Gene Aliases: |
DLG3 |
Location: |
Chr.17:43800799-43833192 on Build GRCh38 |
Assay Gene Location: | Within Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MPP3 | NM_001932.4 | 15 | 1283 | NP_001923.2 |
| NR_003562.2 | 14 | 1262 | ||
| XM_006721915.2 | 16 | 1471 | XP_006721978.1 | |
| XM_011524828.2 | 15 | 1366 | XP_011523130.1 | |
| XM_017024656.1 | 15 | 1499 | XP_016880145.1 | |
| XM_017024657.1 | 14 | 1175 | XP_016880146.1 | |
| AB209232.1 | 1 | 399 | BAD92469.1 | |
| AK311368.1 | 13 | 1359 | ||
| AK313045.1 | 15 | 1250 | ||
| AM050144.1 | 13 | 1114 | CAJ18313.1 | |
| AM050145.1 | 12 | 1093 | CAJ18315.1 | |
| BC047017.1 | 14 | 1253 | AAH47017.1 | |
| BC056865.1 | 14 | 1197 | AAH56865.1 | |
| U37707.1 | 13 | 1444 | AAB36964.1 |
More Information
Set Membership: |
|

Genomic Map