Assay Details
Target Gene Details
Entrez Gene ID: | 6294 |
Gene Name: | scaffold attachment factor B |
Gene Aliases: |
HAP, HET, SAB-B1, SAF-B, SAF-B1, SAFB1 |
Location: |
Chr.19:5623035-5668478 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SAFB | NM_001201338.1 | 1 | 171 | NP_001188267.1 |
| NM_001201339.1 | 1 | 171 | NP_001188268.1 | |
| NM_001201340.1 | 1 | 171 | NP_001188269.1 | |
| NM_001320571.1 | 1 | 171 | NP_001307500.1 | |
| NM_001320572.1 | 1 | 171 | NP_001307501.1 | |
| NM_002967.3 | 1 | 171 | NP_002958.2 | |
| XM_017027114.1 | 1 | 125 | XP_016882603.1 | |
| XM_017027115.1 | 1 | 125 | XP_016882604.1 | |
| XM_017027116.1 | XP_016882605.1 | |||
| XM_017027117.1 | XP_016882606.1 | |||
| XM_017027118.1 | XP_016882607.1 | |||
| AB208780.1 | 1 | 88 | BAD92017.1 | |
| AK095736.1 | ||||
| AK290444.1 | ||||
| AK293152.1 | ||||
| AK294718.1 | 1 | 123 | ||
| AK295298.1 | ||||
| AK298707.1 | ||||
| AK299525.1 | ||||
| AK304474.1 | ||||
| AK307970.1 | ||||
| BC126219.1 | ||||
| BC143937.1 | ||||
| BC143938.1 | ||||
| BC143939.1 | ||||
| DC349269.1 | 1 | 157 | ||
| U72355.1 | AAC00056.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1062851 | Chr.19:5531540 - 5893590 on Build GRCh38 | Gain |
|
| nsv2395 | Chr.19:5613162 - 5644262 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map