Assay Details
Target Gene Details
Entrez Gene ID: | 51130 |
Gene Name: | ankyrin repeat and SOCS box containing 3 |
Gene Aliases: |
ASB-3 |
Location: |
Chr.2:53669980-53787009 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ASB3 | NM_001201965.1 | NP_001188894.1 | ||
| NM_016115.4 | NP_057199.1 | |||
| NM_145863.2 | NP_665862.1 | |||
| AB209795.1 | BAD93032.1 | |||
| AF156778.1 | AAD41895.1 | |||
| AK000985.1 | BAA91455.1 | |||
| AK001283.1 | BAA91599.1 | |||
| AK056069.1 | ||||
| AK222595.1 | BAD96315.1 | |||
| AK222715.1 | BAD96435.1 | |||
| AK303133.1 | ||||
| BC006488.2 | AAH06488.1 | |||
| BC009569.1 | AAH09569.1 |
Target Gene Details
Entrez Gene ID: | 100302652 |
Gene Name: | GPR75-ASB3 readthrough |
Gene Aliases: |
- |
Location: |
Chr.2:53670293-53860033 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GPR75-ASB3 | NM_001164165.1 | NP_001157637.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv3844n100 | Chr.2:53667392 - 53720644 on Build GRCh38 | Loss |
|
| nsv834058 | Chr.2:53673537 - 53844874 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map