Assay Details
Target Gene Details
Entrez Gene ID: | 84897 |
Gene Name: | transforming growth factor beta regulator 1 |
Gene Aliases: |
NIAM, TB-5 |
Location: |
Chr.11:124622599-124635926 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TBRG1 | NM_032811.2 | 1 | 155 | NP_116200.2 |
| NR_016021.1 | 1 | 155 | ||
| XM_005271709.3 | 1 | 402 | XP_005271766.1 | |
| AK027527.1 | 1 | 126 | BAB55177.1 | |
| AK057749.1 | 1 | 126 | BAB71559.1 | |
| AK074594.1 | 1 | 141 | ||
| AK222936.1 | 1 | 131 | BAD96656.1 | |
| BC018452.1 | 1 | 123 | ||
| BC032312.1 | 1 | 97 | AAH32312.1 | |
| BC041043.1 | 1 | 140 | ||
| BC073160.1 | 1 | 84 | AAH73160.1 | |
| BC109269.2 | AAI09270.1 | |||
| DA292015.1 | 1 | 155 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1045156 | Chr.11:124565356 - 124705852 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map