Assay Details
Target Gene Details
Entrez Gene ID: | 51557 |
Gene Name: | lengsin, lens protein with glutamine synthetase domain |
Gene Aliases: |
GLULD1, LGS |
Location: |
Chr.6:63275951-63567921 on Build GRCh38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LGSN | NM_001143940.1 | NP_001137412.1 | ||
| NM_016571.2 | 4 | 768 | NP_057655.2 | |
| XM_011535889.2 | 10 | 1885 | XP_011534191.1 | |
| XM_011535892.2 | 10 | 1741 | XP_011534194.1 | |
| XM_017010929.1 | 11 | 4222 | XP_016866418.1 | |
| XM_017010930.1 | 10 | 1804 | XP_016866419.1 | |
| XM_017010931.1 | 9 | 1552 | XP_016866420.1 | |
| AF242388.1 | 4 | 768 | AAF61255.1 | |
| BC130366.1 | 4 | 767 | ||
| BC130368.1 | 4 | 767 | ||
| DQ682605.1 | ||||
| DQ682606.1 | 5 | 816 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1019831 | Chr.6:62569772 - 63520831 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map