Assay Details
Target Gene Details
Entrez Gene ID: | 84958 |
Gene Name: | synaptotagmin like 1 |
Gene Aliases: |
JFC1, SLP1 |
Location: |
Chr.1:27341992-27353932 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 4 - Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SYTL1 | NM_001193308.1 | NP_001180237.1 | ||
| NM_032872.2 | NP_116261.1 | |||
| XM_005246022.3 | XP_005246079.1 | |||
| XM_006710990.1 | XP_006711053.1 | |||
| XM_006710991.1 | XP_006711054.1 | |||
| AK027902.1 | BAB55444.1 | |||
| AK074154.1 | BAB84980.1 | |||
| AK096437.1 | ||||
| AK097172.1 | BAC04970.1 | |||
| AK293038.1 | ||||
| AY037157.1 | AAK67636.1 | |||
| BC009224.2 | AAH09224.2 | |||
| BC015764.1 | AAH15764.1 | |||
| BC035725.1 | AAH35725.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv526051 | Chr.1:27284255 - 27445496 on Build GRCh38 | Gain |
|
| nsv516766 | Chr.1:27335744 - 27405147 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map