Assay Details
Target Gene Details
Entrez Gene ID: | 101927870 |
Gene Name: | uncharacterized LOC101927870 |
Gene Aliases: |
- |
Location: |
Chr.7:103445207-103514007 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC101927870 | NR_110141.1 |
Target Gene Details
Entrez Gene ID: | 5649 |
Gene Name: | reelin |
Gene Aliases: |
ETL7, LIS2, PRO1598, RL |
Location: |
Chr.7:103471784-103989516 on Build GRCh38 |
Assay Gene Location: | Within Exon 56 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RELN | NM_005045.3 | 56 | 9223 | NP_005036.2 |
| NM_173054.2 | 56 | 9223 | NP_774959.1 | |
| AK293863.1 | 13 | 2159 | ||
| AK297459.1 | 6 | 787 | ||
| U79716.1 | 56 | 9238 | AAC51105.1 |
More Information
Set Membership: |
|

Genomic Map