Assay Details
Target Gene Details
Entrez Gene ID: | 25842 |
Gene Name: | anti-silencing function 1A histone chaperone |
Gene Aliases: |
CGI-98, CIA, HSPC146 |
Location: |
Chr.6:118894077-118909171 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ASF1A | NM_014034.2 | 4 | 1709 | NP_054753.1 |
| AK025738.1 | 4 | 1566 | BAB15228.1 | |
| AL050261.1 | 4 | 1564 | CAB43363.1 |
Target Gene Details
Entrez Gene ID: | 254394 |
Gene Name: | minichromosome maintenance 9 homologous recombination repair factor |
Gene Aliases: |
C6orf61, MCMDC1, ODG4, dJ329L24.1, dJ329L24.3 |
Location: |
Chr.6:118813449-118935162 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MCM9 | NM_017696.2 | NP_060166.2 |
More Information
Set Membership: |
|

Genomic Map