Assay Details
Target Gene Details
Entrez Gene ID: | 8669 |
Gene Name: | eukaryotic translation initiation factor 3 subunit J |
Gene Aliases: |
EIF3S1, eIF3-alpha, eIF3-p35 |
Location: |
Chr.15:44537068-44562803 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EIF3J | NM_001284335.1 | NP_001271264.1 | ||
| NM_001284336.1 | NP_001271265.1 | |||
| NM_003758.3 | NP_003749.2 | |||
| AK002006.1 | ||||
| AK023388.1 | BAB14555.1 | |||
| AK300659.1 | ||||
| AL536561.3 | ||||
| BC002719.2 | AAH02719.1 | |||
| CN414611.1 | ||||
| U97670.1 | AAC78729.1 |
More Information
Set Membership: |
|

Genomic Map