Assay Details
Target Gene Details
Entrez Gene ID: | 83447 |
Gene Name: | solute carrier family 25 member 31 |
Gene Aliases: |
AAC4, ANT 4, ANT4, SFEC35kDa |
Location: |
Chr.4:127730375-127774299 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC25A31 | NM_001318467.1 | 7 | 1089 | NP_001305396.1 |
| NM_031291.3 | 6 | 1037 | NP_112581.1 | |
| XM_011532298.1 | 4 | 767 | XP_011530600.1 | |
| AJ863129.1 | CAI05952.1 | |||
| AL136857.1 | 6 | 996 | CAB66791.1 | |
| AY550240.1 | 6 | 988 | ||
| BC022032.2 | 6 | 988 | AAH22032.1 | |
| BM563946.1 | ||||
| BU853653.1 | 3 | 274 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv830057 | Chr.4:127670133 - 127867622 on Build GRCh38 | Loss |
|
| nsv437935 | Chr.4:127768781 - 127782795 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map