Assay Details
Target Gene Details
Entrez Gene ID: | 152503 |
Gene Name: | SH3 domain containing 19 |
Gene Aliases: |
EBP, EVE1, Kryn, SH3P19 |
Location: |
Chr.4:151120281-151228030 on Build GRCh38 |
Assay Gene Location: | Within Exon 23 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SH3D19 | NM_001009555.3 | 21 | 4760 | NP_001009555.3 |
| NM_001128923.1 | 20 | 4679 | NP_001122395.1 | |
| NM_001128924.1 | 13 | 3682 | NP_001122396.1 | |
| NM_001243349.1 | 20 | 4600 | NP_001230278.1 | |
| XM_005262767.2 | 22 | 4621 | XP_005262824.1 | |
| XM_011531646.1 | 20 | 4407 | XP_011529948.1 | |
| XM_011531647.1 | 19 | 4299 | XP_011529949.1 | |
| XM_011531648.1 | 22 | 4734 | XP_011529950.1 | |
| XM_011531649.1 | 18 | 4230 | XP_011529951.1 | |
| XM_017007778.1 | 22 | 4725 | XP_016863267.1 | |
| XM_017007779.1 | 18 | 4221 | XP_016863268.1 | |
| XM_017007780.1 | 21 | 4656 | XP_016863269.1 | |
| XM_017007781.1 | 21 | 4651 | XP_016863270.1 | |
| XM_017007782.1 | 20 | 4582 | XP_016863271.1 | |
| XM_017007783.1 | 21 | 4552 | XP_016863272.1 | |
| AK128493.1 | 13 | 3531 | ||
| AK294476.1 | 20 | 4600 | ||
| AL133047.1 | 11 | 2524 | CAB61374.1 | |
| BC032468.1 | 3 | 1613 | AAH32468.1 | |
| BC045742.1 | 5 | 1925 | AAH45742.1 | |
| BC085613.1 | 12 | 2739 | ||
| BM670337.1 | ||||
| BM831166.1 | 1 | 292 | ||
| BX647422.1 | 21 | 4672 | CAI46052.1 | |
| CF541231.1 | 1 | 525 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv595690 | Chr.4:150867803 - 151188400 on Build GRCh38 | Gain |
|
| esv3602674 | Chr.4:151094056 - 151193747 on Build GRCh38 | Gain |
|
| dgv5436n100 | Chr.4:150743286 - 151150724 on Build GRCh38 | Gain |
|
| nsv1028814 | Chr.4:150874356 - 152285445 on Build GRCh38 | Gain |
|
| nsv526976 | Chr.4:150754667 - 151133921 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map