Assay Details
Target Gene Details
Entrez Gene ID: | 102465486 |
Gene Name: | microRNA 6811 |
Gene Aliases: |
hsa-mir-6811 |
Location: |
Chr.2:237510931-237510988 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MIR6811 | NR_106869.1 |
Target Gene Details
Entrez Gene ID: | 79083 |
Gene Name: | melanophilin |
Gene Aliases: |
SLAC2-A |
Location: |
Chr.2:237486410-237555318 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MLPH | NM_001042467.2 | NP_001035932.1 | ||
| NM_001281473.1 | NP_001268402.1 | |||
| NM_001281474.1 | NP_001268403.1 | |||
| NM_024101.6 | NP_077006.1 | |||
| NR_104019.1 | ||||
| XM_006712737.1 | XP_006712800.1 | |||
| XM_006712739.1 | XP_006712802.1 | |||
| XM_006712740.1 | XP_006712803.1 | |||
| XM_017004893.1 | XP_016860382.1 | |||
| XM_017004894.1 | XP_016860383.1 | |||
| AK022207.1 | BAB13984.1 | |||
| AK056867.1 | ||||
| AK094168.1 | ||||
| AK225381.1 | ||||
| AK292929.1 | ||||
| AK296745.1 | ||||
| AK310821.1 | ||||
| BC001653.2 | AAH01653.1 | |||
| BC051269.1 | AAH51269.1 | |||
| BM272410.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv584728 | Chr.2:237362083 - 237525791 on Build GRCh38 | Loss |
|
| nsv1000252 | Chr.2:235899999 - 238766417 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs74322818] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map