Assay Details
Target Gene Details
Entrez Gene ID: | 51455 |
Gene Name: | REV1, DNA directed polymerase |
Gene Aliases: |
AIBP80, REV1L |
Location: |
Chr.2:99400475-99490123 on Build GRCh38 |
Assay Gene Location: | Within Exon 28 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| REV1 | NM_001037872.2 | 23 | 4539 | NP_001032961.1 |
| NM_001321454.1 | 24 | 4650 | NP_001308383.1 | |
| NM_001321455.1 | 24 | 4696 | NP_001308384.1 | |
| NM_001321458.1 | 24 | 4666 | NP_001308387.1 | |
| NM_001321459.1 | 24 | 4742 | NP_001308388.1 | |
| NM_001321460.1 | 24 | 4739 | NP_001308389.1 | |
| NM_016316.3 | 23 | 4542 | NP_057400.1 | |
| NR_135649.1 | 23 | 5036 | ||
| NR_135650.1 | 23 | 4993 | ||
| NR_135651.1 | 22 | 4373 | ||
| NR_135652.1 | 22 | 4839 | ||
| NR_135653.1 | 22 | 4376 | ||
| BM683065.1 | 1 | 248 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv524466 | Chr.2:99335623 - 99415089 on Build GRCh38 | Loss |
|
| dgv4032n100 | Chr.2:99364388 - 99481422 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map