Assay Details
Target Gene Details
Entrez Gene ID: | 101929550 |
Gene Name: | uncharacterized LOC101929550 |
Gene Aliases: |
- |
Location: |
Chr.8:35672163-35710320 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC101929550 | NR_125819.1 | |||
| AL137475.1 |
Target Gene Details
Entrez Gene ID: | 137970 |
Gene Name: | unc-5 netrin receptor D |
Gene Aliases: |
PRO34692, Unc5h4 |
Location: |
Chr.8:35235457-35796550 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| UNC5D | NM_001322818.1 | 5 | 662 | NP_001309747.1 |
| NM_080872.3 | 5 | 949 | NP_543148.2 | |
| XM_011544400.2 | 5 | 923 | XP_011542702.1 | |
| XM_011544401.2 | 5 | 923 | XP_011542703.1 | |
| XM_011544402.2 | 5 | 923 | XP_011542704.1 | |
| XM_011544403.2 | 5 | 923 | XP_011542705.1 | |
| XM_011544404.2 | 5 | 923 | XP_011542706.1 | |
| XM_017013044.1 | 5 | 924 | XP_016868533.1 | |
| XM_017013045.1 | 5 | 924 | XP_016868534.1 | |
| XM_017013046.1 | 5 | 924 | XP_016868535.1 | |
| XM_017013047.1 | 5 | 923 | XP_016868536.1 | |
| XM_017013048.1 | 5 | 2109 | XP_016868537.1 | |
| XM_017013049.1 | 5 | 2060 | XP_016868538.1 | |
| XM_017013050.1 | 6 | 1516 | XP_016868539.1 | |
| AB055056.1 | 5 | 661 | BAB83663.1 | |
| AK122615.1 | ||||
| AY358147.1 | 5 | 949 | AAQ88514.1 | |
| BY798317.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831285 | Chr.8:35628356 - 35790599 on Build GRCh38 | Loss |
|
| nsv6152 | Chr.8:35678315 - 35711831 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map