Assay Details
Target Gene Details
Entrez Gene ID: | 84795 |
Gene Name: | pyridine nucleotide-disulphide oxidoreductase domain 2 |
Gene Aliases: |
C10orf33, FP3420 |
Location: |
Chr.10:98383565-98415221 on Build GRCh38 |
Assay Gene Location: | Within Exon 22 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PYROXD2 | NM_032709.2 | 16 | 1900 | NP_116098.2 |
| XM_011540293.2 | 17 | 3322 | XP_011538595.1 | |
| XM_011540301.2 | 9 | 2736 | XP_011538603.1 | |
| XM_017016838.1 | 16 | 4096 | XP_016872327.1 | |
| XM_017016839.1 | 14 | 1953 | XP_016872328.1 | |
| XM_017016840.1 | 14 | 1807 | XP_016872329.1 | |
| XM_017016841.1 | 13 | 2114 | XP_016872330.1 | |
| AF461898.1 | 1 | 2368 | AAQ04812.1 | |
| AK074429.1 | 16 | 1861 | ||
| AK075265.1 | 16 | 1838 | BAC11507.1 | |
| AK123713.1 | 6 | 1857 | ||
| AK225637.1 | 16 | 1854 | ||
| BC006131.1 | 16 | 1863 | AAH06131.1 |
More Information
Set Membership: |
|

Genomic Map