Assay Details
Target Gene Details
Entrez Gene ID: | 7372 |
Gene Name: | uridine monophosphate synthetase |
Gene Aliases: |
OPRT |
Location: |
Chr.3:124730366-124749273 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| UMPS | NM_000373.3 | 2 | 322 | NP_000364.1 |
| NR_033434.1 | ||||
| NR_033437.1 | 3 | 421 | ||
| AB062285.1 | 2 | 244 | BAB93468.1 | |
| AK291306.1 | 2 | 238 | ||
| AL080099.2 | CAB45710.3 | |||
| BC000364.2 | 2 | 239 | AAH00364.1 | |
| BC007511.2 | 2 | 239 | AAH07511.1 | |
| CR456787.1 | 2 | 216 | CAG33068.1 | |
| D86227.1 | 2 | 235 | BAA19920.1 | |
| D86228.1 | 2 | 235 | BAA19921.1 | |
| D86230.1 | 2 | 235 | BAA19923.1 | |
| EU921886.1 | 2 | 236 | ||
| EU921887.1 | ||||
| EU921888.1 | ||||
| EU921889.1 | ||||
| EU921890.1 | 2 | 236 | ||
| EU921891.1 | 3 | 335 | ||
| EU921892.1 | 2 | 236 | ||
| EU921893.1 | 2 | 236 | ||
| EU921894.1 | 2 | 236 | ||
| EU921895.1 | ||||
| J03626.1 | 2 | 320 | AAA61255.1 | |
| KU178375.1 | 2 | 216 | ||
| KU178376.1 | 3 | 315 | ||
| M36661.1 | 2 | 182 | AAA61256.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1012242 | Chr.3:124722848 - 124797029 on Build GRCh38 | Gain |
|
| nsv1001677 | Chr.3:124015843 - 125108694 on Build GRCh38 | Loss |
|
| nsv1160974 | Chr.3:124577493 - 125300995 on Build GRCh38 | Duplication |
|
More Information
Set Membership: |
|

Genomic Map