Assay Details
Target Gene Details
Entrez Gene ID: | 4199 |
Gene Name: | malic enzyme 1 |
Gene Aliases: |
HUMNDME, MES |
Location: |
Chr.6:83210389-83431219 on Build GRCh38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ME1 | NM_002395.5 | 14 | 2646 | NP_002386.1 |
| XM_011535836.2 | 13 | 2371 | XP_011534138.1 | |
| AJ420574.1 | 1 | 498 |
More Information
Set Membership: |
|

Genomic Map