Assay Details
Target Gene Details
Entrez Gene ID: | 120103 |
Gene Name: | solute carrier family 36 member 4 |
Gene Aliases: |
PAT4 |
Location: |
Chr.11:93144171-93197983 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC36A4 | NM_001286139.1 | NP_001273068.1 | ||
| NM_152313.3 | NP_689526.2 | |||
| XM_017017174.1 | XP_016872663.1 | |||
| XM_017017175.1 | XP_016872664.1 | |||
| XM_017017176.1 | XP_016872665.1 | |||
| XM_017017177.1 | XP_016872666.1 | |||
| AK096251.1 | BAC04737.1 | |||
| AY162216.1 | AAO11790.1 | |||
| BC047374.1 | AAH47374.1 | |||
| HY170772.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1039134 | Chr.11:93056834 - 93486776 on Build GRCh38 | Loss |
|
| nsv818858 | Chr.11:93165176 - 93199063 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map