Assay Details
Target Gene Details
Entrez Gene ID: | 23635 |
Gene Name: | single stranded DNA binding protein 2 |
Gene Aliases: |
HSPC116, SOSS-B2 |
Location: |
Chr.5:81417360-81751253 on Build GRCh38 |
Assay Gene Location: | Within Exon 21 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SSBP2 | NM_001256732.1 | 17 | 1366 | NP_001243661.1 |
| NM_001256733.1 | 16 | 1282 | NP_001243662.1 | |
| NM_001256734.1 | 16 | 1276 | NP_001243663.1 | |
| NM_001256735.1 | 16 | 1252 | NP_001243664.1 | |
| NM_001256736.1 | 15 | 1153 | NP_001243665.1 | |
| NM_012446.3 | 17 | 1342 | NP_036578.2 | |
| XM_011543297.2 | 16 | 1182 | XP_011541599.1 | |
| XM_017009304.1 | 17 | 1184 | XP_016864793.1 | |
| XM_017009305.1 | 18 | 1273 | XP_016864794.1 | |
| XM_017009306.1 | 16 | 1158 | XP_016864795.1 | |
| XM_017009307.1 | 16 | 1070 | XP_016864796.1 | |
| XM_017009308.1 | 18 | 1445 | XP_016864797.1 | |
| XM_017009309.1 | 17 | 1509 | XP_016864798.1 | |
| XM_017009310.1 | 17 | 1657 | XP_016864799.1 | |
| AF077048.1 | AAD27781.1 | |||
| AF161465.1 | 17 | 1172 | AAF29080.1 | |
| AK074537.1 | 17 | 1137 | BAC11046.1 | |
| AK293544.1 | 16 | 1077 | ||
| AK294864.1 | 15 | 948 | ||
| AK297494.1 | 17 | 1050 | ||
| AK299858.1 | 16 | 1120 | ||
| AL080076.1 | 17 | 1190 | CAB45699.1 | |
| BC017020.1 | 17 | 1190 | AAH17020.1 | |
| BX397789.2 | 1 | 557 | ||
| BX413405.2 | 1 | 544 | ||
| CR936787.1 | 16 | 1097 | ||
| DA202987.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3605594 | Chr.5:81406976 - 81473957 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map