Assay Details
Target Gene Details
Entrez Gene ID: | 221711 |
Gene Name: | synaptonemal complex protein 2 like |
Gene Aliases: |
C6orf177, NO145, dJ62D2.1 |
Location: |
Chr.6:10886831-10974309 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 14 - Intron 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SYCP2L | NM_001040274.2 | NP_001035364.2 | ||
| AK128130.1 | BAC87288.1 | |||
| AM293674.1 | ||||
| BC012225.1 | AAH12225.1 | |||
| DA620756.1 |
More Information
Set Membership: |
|

Genomic Map