Assay Details
Target Gene Details
Entrez Gene ID: | 8848 |
Gene Name: | TSC22 domain family member 1 |
Gene Aliases: |
Ptg-2, TGFB1I4, TSC22 |
Location: |
Chr.13:44432143-44576565 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TSC22D1 | NM_001243799.1 | 1 | 305 | NP_001230728.1 |
| NM_183422.3 | 1 | 305 | NP_904358.2 | |
| XM_005266583.4 | 1 | 168 | XP_005266640.1 | |
| XM_017020810.1 | 1 | 168 | XP_016876299.1 | |
| XM_017020811.1 | 1 | 168 | XP_016876300.1 | |
| AB082525.1 | 1 | 305 | ||
| AK091854.1 | ||||
| BC136475.1 | 1 | 75 | ||
| BC136482.1 | 1 | 75 |
Target Gene Details
Entrez Gene ID: | 641467 |
Gene Name: | TSC22D1 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.13:44575896-44580432 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TSC22D1-AS1 | NR_038381.1 |
More Information
Set Membership: |
|

Genomic Map