Assay Details
Target Gene Details
Entrez Gene ID: | 11226 |
Gene Name: | polypeptide N-acetylgalactosaminyltransferase 6 |
Gene Aliases: |
GALNAC-T6, GalNAcT6 |
Location: |
Chr.12:51351870-51391955 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GALNT6 | NM_007210.3 | 1 | 41 | NP_009141.2 |
| XM_005268607.1 | XP_005268664.1 | |||
| XM_006719214.2 | XP_006719277.1 | |||
| XM_011537822.2 | XP_011536124.1 | |||
| XM_017018745.1 | XP_016874234.1 | |||
| AK296852.1 | ||||
| BC035822.1 | 1 | 44 | AAH35822.2 | |
| DB001644.1 | 1 | 216 |
Target Gene Details
Entrez Gene ID: | 9498 |
Gene Name: | solute carrier family 4 member 8 |
Gene Aliases: |
NBC3, NDCBE |
Location: |
Chr.12:51391317-51515763 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC4A8 | NM_001258401.2 | NP_001245330.1 | ||
| NM_001267615.1 | NP_001254544.1 | |||
| XM_011539011.1 | XP_011537313.1 | |||
| AB018282.1 | BAA34459.1 | |||
| AK295315.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832408 | Chr.12:51302821 - 51477555 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs115491823] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map