Assay Details
Target Gene Details
Entrez Gene ID: | 221184 |
Gene Name: | copine 2 |
Gene Aliases: |
COPN2, CPN2 |
Location: |
Chr.16:57092543-57147966 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CPNE2 | NM_152727.5 | 3 | 581 | NP_689940.3 |
| AF492484.1 | 3 | 532 | AAO85483.1 | |
| AK292269.1 | 2 | 490 | ||
| AK315394.1 | 3 | 484 | ||
| BC053658.1 | 4 | 655 | AAH53658.1 | |
| BG830959.1 | 3 | 527 | ||
| BG831872.1 | 3 | 529 | ||
| CR749617.1 | 2 | 242 | CAH18411.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv827683 | Chr.16:57016822 - 57147283 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map