Assay Details
Target Gene Details
Entrez Gene ID: | 2571 |
Gene Name: | glutamate decarboxylase 1 |
Gene Aliases: |
CPSQ1, GAD, SCP |
Location: |
Chr.2:170813210-170861151 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GAD1 | NM_000817.2 | NP_000808.2 | ||
| NM_013445.3 | NP_038473.2 | |||
| XM_005246444.2 | XP_005246501.1 | |||
| XM_011510922.1 | XP_011509224.1 | |||
| XM_017003757.1 | XP_016859246.1 | |||
| XM_017003758.1 | XP_016859247.1 | |||
| BC037780.2 | AAH37780.1 | |||
| M81883.1 | 1 | 141 | AAA62368.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv528929 | Chr.2:170815681 - 170821869 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map