Assay Details
Target Gene Details
Entrez Gene ID: | 417 |
Gene Name: | ADP-ribosyltransferase 1 |
Gene Aliases: |
ART2, ARTC1, CD296, RT6 |
Location: |
Chr.11:3645110-3665595 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ART1 | XM_011520114.2 | XP_011518416.1 | ||
| XM_017017763.1 | XP_016873252.1 |
Target Gene Details
Entrez Gene ID: | 57053 |
Gene Name: | cholinergic receptor nicotinic alpha 10 subunit |
Gene Aliases: |
- |
Location: |
Chr.11:3665587-3673208 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CHRNA10 | NM_001303034.1 | NP_001289963.1 | ||
| NM_001303035.1 | NP_001289964.1 | |||
| NM_020402.3 | NP_065135.2 | |||
| XM_011520234.1 | XP_011518536.1 | |||
| AF199235.2 | AAG00795.2 | |||
| AJ278118.1 | CAC20435.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2759796 | Chr.11:3215282 - 3780199 on Build GRCh38 | Gain+Loss |
|
| nsv832052 | Chr.11:3595654 - 3780199 on Build GRCh38 | Loss |
|
| nsv526441 | Chr.11:3604947 - 3735456 on Build GRCh38 | Gain |
|
| nsv832051 | Chr.11:3502717 - 3707484 on Build GRCh38 | Gain |
|
| nsv1046356 | Chr.11:3607607 - 3752985 on Build GRCh38 | Gain |
|
| esv3625147 | Chr.11:3607625 - 3732043 on Build GRCh38 | Gain |
|
| nsv482156 | Chr.11:3665587 - 3671384 on Build GRCh38 | Loss |
|
| esv3625148 | Chr.11:3609614 - 3680474 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map