Assay Details
Target Gene Details
Entrez Gene ID: | 56479 |
Gene Name: | potassium voltage-gated channel subfamily Q member 5 |
Gene Aliases: |
Kv7.5 |
Location: |
Chr.6:72621843-73198851 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KCNQ5 | NM_001160130.1 | 1 | 293 | NP_001153602.1 |
| NM_001160132.1 | 1 | 293 | NP_001153604.1 | |
| NM_001160133.1 | 1 | 293 | NP_001153605.1 | |
| NM_001160134.1 | 1 | 293 | NP_001153606.1 | |
| NM_019842.3 | 1 | 293 | NP_062816.2 | |
| XM_017011058.1 | 1 | 293 | XP_016866547.1 | |
| AF249278.1 | 1 | 55 | AAF91335.1 | |
| BC050689.1 | AAH50689.1 | |||
| BC117359.1 | ||||
| BC143554.1 | ||||
| DA801374.1 | 1 | 293 |
More Information
Set Membership: |
|

Genomic Map