Assay Details
Target Gene Details
Entrez Gene ID: | 770 |
Gene Name: | carbonic anhydrase 11 |
Gene Aliases: |
CARPX1 |
Location: |
Chr.19:48637942-48646312 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CA11 | NM_001217.4 | NP_001208.2 | ||
| NR_136241.1 | ||||
| AB018195.1 | BAA36840.1 | |||
| AF050106.1 | AAD08802.1 | |||
| AF067662.1 | AAC99689.1 | |||
| AY358967.1 | AAQ89326.1 | |||
| BC002662.2 | AAH02662.1 | |||
| BT007265.1 | AAP35929.1 | |||
| CR541772.1 | CAG46571.1 |
Target Gene Details
Entrez Gene ID: | 653677 |
Gene Name: | secretory blood group 1, pseudogene |
Gene Aliases: |
SEC1 |
Location: |
Chr.19:48638039-48682245 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SEC1P | NR_004401.2 | |||
| AK301046.1 | ||||
| AK301238.1 | ||||
| D87946.1 | ||||
| D87947.1 | ||||
| DC353776.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv9739 | Chr.19:47901840 - 50143063 on Build GRCh38 | Gain+Loss |
|
| dgv3617n100 | Chr.19:48532114 - 48643732 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map